Learn More
Macherey-Nagel™ Nucleoshell™ RP 18 C18 Reversed Phase HPLC Column, 2.7 μm, 3 mm x 100 mm
Column Format: Analytical Column; Carbon Load: 7.80%; Pore Size: 90 Ă…; Column Phase: Reversed Phase
Brand: Macherey-Nagel™ 763134.30
This item is not returnable.
View return policy
Specifications
Analytical Column | |
3 mm | |
100 mm | |
2.7 ÎĽm | |
90 Ă… | |
Acetonitrile/water | |
60°C | |
C18 | |
7.8% | |
Yes | |
EC Column |
Octadecyl | |
For LC/MS and HPLC | |
8700 psi (600 bar) | |
1 to 11 | |
1 Ea. | |
130 m²/g | |
L1 | |
Silica | |
Reversed Phase | |
Reversed Phase | |
HPLC, LC/MS |
Product Content Correction
Product Title
Macherey-Nagel™ Nucleoshell™ RP 18 C18 Reversed Phase HPLC Column, 2.7 μm, 3 mm x 100 mm > I.D.: 3mm; Length: 100mm
Spot an opportunity for improvement?Share a Content Correction