Learn More
Thermo Scientific™ Nunc™ Petri Dishes
Prepare samples in hospital environments, food science or the pharmaceutical industry with these petri dishes
2305.00 SEK - 2982.48 SEK
Specifications
Bottom Shape | Flat |
---|---|
Material | Polystyrene |
Shape | Round |
Sterility | Sterile |
Type | Petri Dish |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10051622
|
Thermo Scientific™
249964 |
140 mm |
2305.00 SEK
Pack of 80 |
In Stock
Log in to see stock. |
|||||
10444912
|
Thermo Scientific™
263991 |
90 mm |
2982.48 SEK
Pack of 320 |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
Description
Prepare samples in hospital environments, food science or the pharmaceutical industry with Thermo Scientific™ Nunc™ Petri Dishes. These dishes are available with a molded grid and perform well in automatic dispensers due to complete flatness and uniform height.
- For the culturing of bacteria, fungi and other microorganisms
- Constructed of high-quality and optically clear polystyrene, with lid
- Fitted with top and bottom stacking rings for easier handing and stable stacking
- Vented for gaseous exchange
- Sterilized by gamma irradiation
Specifications
Flat | |
Round | |
Petri Dish |
Polystyrene | |
Sterile | |
1 |