missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP155 Mouse anti-Human, Rat, Clone: 1A7F2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66359-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a centrosomal and microtubule-binding protein which is predicted to have two coiled-coil domains and a rhodanese domain. In human retinal pigment epithelial cells the protein localized to centrioles and cilia. Mutations in this gene have been associated with Joubert Syndrome 15; an autosomal recessive ciliopathy and neurological disorder. Alternative splicing results in multiple transcript variants.Specifications
NUP155 | |
Monoclonal | |
1.5 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75694, P37199 | |
NUP155 | |
NUP155 Fusion Protein Ag3638 | |
20 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG2a |
Western Blot, Immunohistochemistry (Paraffin) | |
1A7F2 | |
Unconjugated | |
NUP155 | |
155 kDa nucleoporin, KIAA0791, N155, nucleoporin 155kDa, Nucleoporin Nup155, NUP155 | |
Mouse | |
Protein A | |
RUO | |
117021, 9631 | |
-20°C | |
Liquid |