missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP155 Mouse anti-Human, Rat, Clone: 1A7F2, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | NUP155 |
---|---|
Clone | 1A7F2 |
Concentration | 1.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16828165
|
Proteintech
66359-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16818165
|
Proteintech
66359-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a centrosomal and microtubule-binding protein which is predicted to have two coiled-coil domains and a rhodanese domain. In human retinal pigment epithelial cells the protein localized to centrioles and cilia. Mutations in this gene have been associated with Joubert Syndrome 15; an autosomal recessive ciliopathy and neurological disorder. Alternative splicing results in multiple transcript variants.Specifications
NUP155 | |
1.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
155 kDa nucleoporin, KIAA0791, N155, nucleoporin 155kDa, Nucleoporin Nup155, NUP155 | |
NUP155 | |
IgG2a | |
Protein A | |
Antibody |
1A7F2 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Rat | |
O75694, P37199 | |
117021, 9631 | |
NUP155 Fusion Protein Ag3638 | |
Primary | |
-20°C | |
NUP155 |