Learn More
NUP160 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16084-1-AP-150UL
 View more versions of this product
Product Code. 16885564
  • 4652.90 SEK / 150µL
Description
NUP160 is 1 of up to 60 proteins that make up the 120-MD nuclear pore complex, which mediates nucleoplasmic transport.
Specifications
Specifications
NUP160
Polyclonal
Unconjugated
NUP160
160 kDa nucleoporin, KIAA0197, nucleoporin 160kDa, Nucleoporin Nup160, NUP120, NUP160
Rabbit
Antigen Affinity Chromatography
RUO
23279, 311182, 59015
-20°C
Liquid
Immunoprecipitation, Western Blot
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q12769, Q9Z0W3
Nup160
NUP160 Fusion Protein Ag9083
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
NUP160 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated