missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP62 Mouse anti-Human, Mouse, Rat, Clone: 2G12B3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66573-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
SDS3 is a subunit of the histone deacetylase.Specifications
NUP62 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P17955, P37198, Q63850 | |
Nup62 | |
NUP62 Fusion Protein Ag4912 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin) | |
2G12B3 | |
Unconjugated | |
NUP62 | |
62 kDa nucleoporin, IBSN, Nuclear pore glycoprotein p62, nucleoporin 62kDa, Nucleoporin Nup62, NUP62, p62, SNDI | |
Mouse | |
Protein G | |
RUO | |
18226, 23636, 65274 | |
-20°C | |
Liquid |