missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP62 Mouse anti-Human, Mouse, Rat, Clone: 2G12B3, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | NUP62 |
---|---|
Clone | 2G12B3 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16828575
|
Proteintech
66573-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16818575
|
Proteintech
66573-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
SDS3 is a subunit of the histone deacetylase.Specifications
NUP62 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
62 kDa nucleoporin, IBSN, Nuclear pore glycoprotein p62, nucleoporin 62kDa, Nucleoporin Nup62, NUP62, p62, SNDI | |
Nup62 | |
IgG1 | |
Protein G | |
Antibody |
2G12B3 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P17955, P37198, Q63850 | |
18226, 23636, 65274 | |
NUP62 Fusion Protein Ag4912 | |
Primary | |
-20°C | |
NUP62 |