Learn More
NUP98-NUP96 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12329-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody was raised against the carboxy-terminal region of Nup98-Nup96, it recognizes all Nup98-Nup96 proteins except Nup98-N. Since the autoproteolysis is very rapid this antibody usually detected 98-115 kDa Nup96 protein in multiple cell lines. It
ZFP64 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 9 C2H2-type zinc fingers. ZFP64 may be involved in transcriptional regulation.Specifications
NUP98-NUP96 | |
Polyclonal | |
Unconjugated | |
NUP98 | |
96 kDa nucleoporin, ADAR2, ADIR2, Nucleoporin Nup96, NUP196, NUP96, NUP98 NUP96, NUP98-NUP96 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
269966, 4928, 81738 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P49793, P52948, Q6PFD9 | |
NUP98 | |
NUP98-NUP96 Fusion Protein Ag3016 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |