missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nurr1/NR4A2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10975-2-AP-20UL
This item is not returnable.
View return policy
Description
The Nuclear receptor related 1 protein (Nurr1) also known as NR4A2 (nuclear receptor subfamily 4, group A, member 2) is a protein that in humans is encoded by the NR4A2 gene. NURR1 is a member of the nuclear receptor family of intracellular transcription factors.Nurr1 plays a key role in the maintece of the dopaminergic system of the brain. Mutations in this gene have been associated with disorders related to dopaminergic dysfunction, including Parkinson′s disease, schizophrenia, and manic depression. Misregulation of this gene may be associated with rheumatoid arthritis. Four transcript variants encoding four distinct isoforms have been identified for this gene. Additional alternate splice variants may exist, but their full-length nature has not been determined.Specifications
Nurr1/NR4A2 | |
Polyclonal | |
Unconjugated | |
NR4A2 | |
HZF 3, NOT, NR4A2, NURR1, Nurr1/NR4A2, Orphan nuclear receptor NURR1, RNR1, TINUR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4929 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P43354 | |
NR4A2 | |
Nurr1/NR4A2 Fusion Protein Ag1418 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Nurr1/NR4A2 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction