Learn More
NYAP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25108-1-AP-20UL
 View more versions of this product
Product Code. 16883645
  • 1645.54 SEK / 20µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
Activates PI3K and concomitantly recruits the WAVE1 complex to the close vicinity of PI3K and regulates neuronal morphogenesis.
Specifications
Specifications
NYAP2
Polyclonal
Unconjugated
NYAP2
KIAA1486, KIAA1486 protein, NYAP2
Rabbit
Antigen Affinity Chromatography
RUO
241134, 57624
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.45 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8BM65, Q9P242
NYAP2
NYAP2 Fusion Protein Ag19004
20 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NYAP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated