Learn More
Fisherbrand™ Achromatic Objective
Ergonomically designed for everyday use with easily accessible controls for stage movement coarse and fine focus plus varying light intensity.
Brand:  Fisherbrand™
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12904321
  • 1390.00 SEK / Each
Estimated Shipment: 18-07-2024
to see stock.
Add to basket
Description
Description
  • For use with Max II Compound Microscopes
  • Monocular and binocular
Specifications
Specifications
Achromat Objective
Max II Compound Microscopes
20X
1 Ea.