Learn More
Fisherbrand™ Achromatic Objective
Ergonomically designed for everyday use with easily accessible controls for stage movement coarse and fine focus plus varying light intensity.
740.00 SEK - 2210.00 SEK
Specifications
Type | Achromat Objective |
---|---|
For Use With (Equipment) | Max II Compound Microscopes |
Quantity | 1 Ea. |
Product Code | Brand | Magnification Power | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Magnification Power | Price | Quantity & Availability | |||||
12994311
|
Fisherbrand™
|
10X |
1005.00 SEK
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
12904321
|
Fisherbrand™
|
20X |
1390.00 SEK
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
12964411
|
Fisherbrand™
|
40X |
1390.00 SEK
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
12984311
|
Fisherbrand™
|
4X |
740.00 SEK
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
12914321
|
Fisherbrand™
|
60X |
2210.00 SEK
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
Description
- For use with Max II Compound Microscopes
- Monocular and binocular
Specifications
Achromat Objective | |
Max II Compound Microscopes | |
1 Ea. |