missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oct3/4 Mouse, Clone: 40/Oct-3, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 565644
2615.08 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Oct3/4 Monoclonal antibody specifically detects Oct3/4 in Human, Mouse samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications
Oct3/4 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Cell Biology | |
Human, Mouse | |
Purified |
Flow Cytometry, Flow Cytometry (Intracellular Staining) | |
40/Oct-3 | |
Brilliant Violet 421 | |
Oct3, OTF3, Oct4, OTF4, POU5F1 | |
Mouse Oct3 aa. 252-372 Recombinant Protein | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Oct3/4 Mouse, Clone: 40/Oct-3, BD Horizon™ > 50μg; BV421
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction