Learn More
n-Octatriacontane, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep S-2886
Quantity | 1 mL |
---|
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
n-Octatriacontane | 7194-85-6 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
2mL amber prescored glass ampule | |
0.1, 99.9 | |
0.1, 99.9 | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC | |
Hexane | |
23599 | |
n-Octatriacontane |
7194-85-6, 67-56-1 | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
C38H78 | |
BVKCQBBZBGYNOP-UHFFFAOYSA-N | |
octatriacontane | |
535.042 | |
1 mL |
Safety and Handling
- n-Octatriacontane
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361-Suspected of damaging fertility or the unborn child.
- H373-May cause damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : n-hexane
Your input is important to us. Please complete this form to provide feedback related to the content on this product.