Learn More
ODC1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28728-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The ODC1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ODC1. This antibody recognizes human, mouse antigen. The ODC1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
ODC1 | |
Polyclonal | |
Unconjugated | |
150 μL | |
4953 | |
IgG |
Western Blot, Immunohistochemistry | |
233 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |