Learn More
OGDH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15212-1-AP-20UL
 View more versions of this product
Product Code. 16894294
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
This antibody is immunized with the C-terminal and can recognize the isoform 1 and 2 with the MW of 116 kDa.
This gene encodes one subunit of the 2-oxoglutarate dehydrogenase complex. This complex catalyzes the overall conversion of 2-oxoglutarate (alpha-ketoglutarate) to succinyl-CoA and CO(2) during the Krebs cycle. The protein is located in the mitochondrial matrix and uses thiamine pyrophosphate as a cofactor. A congenital deficiency in 2-oxoglutarate dehydrogenase activity is believed to lead to hypotonia, metabolic acidosis, and hyperlactatemia.
Specifications
Specifications
OGDH
Polyclonal
Unconjugated
OGDH
AKGDH, E1k, OGDC, OGDC E1, OGDH
Rabbit
Antigen Affinity Chromatography
RUO
18293, 360975, 4967
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q02218, Q5XI78, Q60597
Ogdh
OGDH Fusion Protein Ag7368
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
OGDH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated