Learn More
OGDH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15212-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is immunized with the C-terminal and can recognize the isoform 1 and 2 with the MW of 116 kDa.
This gene encodes one subunit of the 2-oxoglutarate dehydrogenase complex. This complex catalyzes the overall conversion of 2-oxoglutarate (alpha-ketoglutarate) to succinyl-CoA and CO(2) during the Krebs cycle. The protein is located in the mitochondrial matrix and uses thiamine pyrophosphate as a cofactor. A congenital deficiency in 2-oxoglutarate dehydrogenase activity is believed to lead to hypotonia, metabolic acidosis, and hyperlactatemia.Specifications
OGDH | |
Polyclonal | |
Unconjugated | |
OGDH | |
AKGDH, E1k, OGDC, OGDC E1, OGDH | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18293, 360975, 4967 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q02218, Q5XI78, Q60597 | |
Ogdh | |
OGDH Fusion Protein Ag7368 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Product Title
OGDH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction