Learn More
Kern Sohn OIML Class E2 Compact Test Weight
Single test weight constructed of stainless steel.
Brand:  Kern Sohn 316-04
Encompass_Preferred
 View more versions of this product
Product Code. 17332003
  • 516.72 SEK / Each
Only null left
Estimated Shipment: 23-07-2025
Description
  • Compact shape
  • Polished stainless steel
Specifications
Specifications
E2
Stainless Steel
10 g
± 0.06 mg
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Kern Sohn OIML Class E2 Compact Test Weight