Learn More
Kern Sohn OIML Class F1 Rectangular Test Weight
Single test weight constructed of stainless steel.
Brand: Kern Sohn 326-38
This item is not returnable.
View return policy
Description
- Rectangular shape
- Polished stainless steel
Specifications
F1 | |
Stainless Steel | |
20 kg | |
± 100 mg |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Kern Sohn OIML Class F1 Rectangular Test Weight
Spot an opportunity for improvement?Share a Content Correction