Learn More
Ohaus™ OIML E2 Weight Set, NVLAP Certificate
Supplied with a NVLAP Accredited Certificate and Adjusted to Meet or Exceed OIML E2 Tolerances
Brand: Ohaus™ 80780381
Additional Details : Weight : 0.00110kg
Description
- Weight Sets in masses ranging from 1 gram to 20 kilograms are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
- NVLAP Calibration Certificate is accredited to ISO 17025 which is recognizable through ILAC
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
NVLAP Accredited Certificate | |
0.050 kg | |
Protective Case |
Stainless Steel, Aluminum | |
500 mg to 1 mg | |
12/Pk. |