missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OIP5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | OIP5 |
---|---|
Concentration | 0.17 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16899673
|
Proteintech
12142-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16889673
|
Proteintech
12142-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Required for recruitment of CENPA to centromeres and normal chromosome segregation during mitosis.Specifications
OIP5 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
O43482 | |
11339 | |
OIP5 Fusion Protein Ag2790 | |
Primary | |
-20°C | |
OIP5 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
5730547N13Rik, Cancer/testis antigen 86, CT86, LINT 25, MIS18B, MIS18beta, OIP 5, OIP5, Opa interacting protein 5, Protein Mis18 beta | |
OIP5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
OIP5 Rabbit anti-Human, Polyclonal, Proteintech