Learn More
Diba Omnifit™ Packing Sleeves
Sleeves to connect glass columns
Brand:  Diba 006PS-06
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 15264509
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • For Omnifit glass low/medium pressure liquid chromatography columns
  • Offered in multiple I.D. (bore) sizes
Specifications
Specifications
Packing Sleeves
6.6 mm (Bore Size) Columns