Learn More
Kolmi Hopen™ Op-Air™ Type II Printed Medical Masks with Earloops
Click to view available options
Color:
Blue Geometric Print
Dinosaur Print
Olive Branch Print
Unit Size:
Pack of 60
Description
- Protects environment from droplets emitted by wearer
- Feature exclusive Origami folding and Smile welding for better comfort and perfect fit
- Softex inner layer is soft and non-irritating for maximum comfort
- Won't wear out, even during prolonged use
- Intended for healthcare professionals
- May also be worn by patients to reduce risk of spreading infection
Specifications
Specifications
Color | Blue Geometric Print |
Material | Polypropylene |
Dimensions (L x W) | 175 x 88 mm |
Certifications/Compliance (Europe) | (EU) 2017/745, EN 14683:2019+AC:2019, ISO 9001, ISO 13485 |
Disposable | Single-use |
Latex Free | Yes |
Packaging Quantity | 60 Pk. |
Product Content Correction
Product Title
Kolmi Hopen™ Op-Air™ Type II Printed Medical Masks with Earloops >
Spot an opportunity for improvement?Share a Content Correction