Learn More
Kolmi Hopen™ Op-Air™ Type II Printed Medical Masks with Earloops Product Code.: 17790219
In Stock
Only null left
Add to basket
Kolmi Hopen™ Op-Air™ Type II Printed Medical Masks with Earloops
Product Code. 17790219
Dinosaur Print, Pack of 60
Click to view available options
Color:
Blue Geometric Print
Dinosaur Print
Olive Branch Print
Unit Size:
Pack of 60
Product Code. 17790219
Brand: Kolmi Hopen™ M76102DINO
Only null left
In Stock
Add to basket
Only null left
Add to basket
These medical masks come in unique designs and have a soft inner layer for those with sensitive skin.
  • Protects environment from droplets emitted by wearer
  • Feature exclusive Origami folding and Smile welding for better comfort and perfect fit
  • Softex inner layer is soft and non-irritating for maximum comfort
  • Won't wear out, even during prolonged use
  • Intended for healthcare professionals
  • May also be worn by patients to reduce risk of spreading infection
Specifications
Color Dinosaur Print
Material Polypropylene
Dimensions (L x W) 175 x 88 mm
Certifications/Compliance (Europe) (EU) 2017/745, EN 14683:2019+AC:2019, ISO 9001, ISO 13485
Disposable Single-use
Latex Free Yes
Packaging Quantity 60 Pk.
Product Content Correction
Product Title
Kolmi Hopen™ Op-Air™ Type II Printed Medical Masks with Earloops >