Learn More
OPRM1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27625-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16875735
  • 4721.67 SEK / 150µL
Estimated Shipment: 19-04-2024
to see stock.
Description
Description
The OPRM1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human OPRM1. This antibody recognizes human, mouse, rat antigen. The OPRM1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
OPRM1
Polyclonal
Unconjugated
150 μL
4988
IgG
Western Blot, Immunohistochemistry, ELISA
267 μg/mL
Rabbit
Primary
Human, Rat, Mouse