Learn More
Fisherbrand™ ORP Electrodes
For measuring oxidation-reduction potential. Fisherbrand™ ORP Electrodes are available for any connector type.
Brand: Fisherbrand™
Additional Details : Weight : 0.10000kg
Description
- Body manufactured from plastic
- Temperature range of 0° to 80°C
- Gel electrolyte
- pH range of ± 1500 mV
Specifications
S7 | |
Platinum Ring | |
ORP Gelled | |
0°C to 80°C | |
1 m |
Plastic | |
Recommended for low or high temperature measurements (–5 to 100°C) or during rapid temperature variation | |
±1500 | |
120 mm | |
12 mm |