Learn More
OSGEPL1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25694-1-AP-20UL
 View more versions of this product
Product Code. 16804265
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
OSGEPL1, also known as Qri7, is a 414 amino acid protein that belongs to the KAE1/YgjD family and exists as 3 alternatively spliced isoforms. In tRNAs that have codons beginning with adenine, OSGEPL1 is required for the formation of a threonylcarbamoyl group on adenosine. The gene that encodes OSGEPL1 contains about 16,568 bases and maps to human chromosome 2q32.2. Consisting of 237 million bases, chromosome 2 encodes over 1,400 genes and makes up approximately 8% of the human genome. A number of genetic diseases are linked to genes on chromosome 2. Harlequin icthyosis, a rare and morbid skin deformity, is associated with mutations in the ABCA12 gene. The lipid metabolic disorder sitosterolemia is associated with ABCG5 and ABCG8. An extremely rare recessive genetic disorder, Alstrom syndrome, is due to mutations in the ALMS1 gene
Specifications
Specifications
OSGEPL1
Polyclonal
Unconjugated
OSGEPL1
OSGEPL1
Rabbit
Antigen Affinity Chromatography
RUO
64172, 72085
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.26 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6PEB4, Q9H4B0
OSGEPL1
OSGEPL1 Fusion Protein Ag22204
20 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
OSGEPL1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated