missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTOR Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18132-1-AP-20UL
Additional Details : Weight : 0.00010kg
Specifications
OTOR | |
Polyclonal | |
Unconjugated | |
OTOR | |
FDP, Fibrocyte derived protein, MIAL, MIAL1, OTOR, otoraplin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56914 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NRC9 | |
OTOR | |
OTOR Fusion Protein Ag12714 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |