Learn More
OTOR Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18132-1-AP-150UL
 View more versions of this product
Product Code. 16888174
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Specifications
OTOR
Polyclonal
Unconjugated
OTOR
FDP, Fibrocyte derived protein, MIAL, MIAL1, OTOR, otoraplin
Rabbit
Antigen Affinity Chromatography
RUO
56914
-20°C
Liquid
Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9NRC9
OTOR
OTOR Fusion Protein Ag12714
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
OTOR Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated