Learn More
Thermo Scientific™ Oxoid™ Fosfomycin Antimicrobial Susceptibility Disks, 50 μg
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing disks.
Brand: Thermo Scientific™ CT0183B
234.68 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Fosfomycin Antimicrobial Susceptibility Disks.
Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.Disk diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
- Inoculum level
- Presence of contamination
- Resistant mutants
- Beta-lactamase activity
- Antagonism and synergy between antibiotics
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Fosfomycin | |
50ÎĽg | |
FOS | |
Antimicrobial Susceptibility | |
50 Disks/Cartridge |
Complies with SFM guidelines | |
Oxoid Fosfomycin Antimicrobial Susceptibility Disks | |
Antimicrobial Susceptibility Discs | |
5 Cartridges/Pk. | |
50 Discs/Cartridge |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6208564 | Certificate of Analysis | 09/01/2025 | CT0183B |
6191409 | Certificate of Analysis | 21/11/2024 | CT0183B |
6116013 | Certificate of Analysis | 13/06/2024 | CT0183B |
3787168 | Certificate of Analysis | 07/02/2024 | CT0183B |
3731512 | Certificate of Analysis | 28/09/2023 | CT0183B |