Learn More
Fisherbrand™ Oxygen/Temp Probes
Dissolved Oxygen and BOD Probe ideal for use in most laboratory and scientific environments.
Brand:  Fisherbrand™ 023ng25-50BSPV10
2453.13 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11732009
  • 2730.00 SEK / Each
Only null left
Estimated Shipment: 04-09-2025
Add to basket
Description
Description
  • Oxygen/temp probe with 15m cable
  • Large digital display
Specifications
Specifications
Oxygen/Temp probes
15
11T
DO and BOD
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Oxygen/Temp Probes > Cable Size: 15m