Learn More
P glycoprotein Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22336-1-AP-150UL
 View more versions of this product
Product Code. 16881865
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is an ATP-dependent drug efflux pump for xenobiotic compounds with broad substrate specificity. It is responsible for decreased drug accumulation in multidrug-resistant cells and often mediates the development of resistance to anticancer drugs. This protein also functions as a transporter in the blood-brain barrier.
Specifications
Specifications
P glycoprotein
Polyclonal
Unconjugated
ABCB1
ABCB1, CD243, GP170, MDR1, Multidrug resistance protein 1, P glycoprotein 1
Rabbit
Antigen Affinity Chromatography
RUO
18669, 18671, 5243
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.48 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P06795, P08183, P21447
ABCB1, Abcb1a, Abcb1b
P glycoprotein Fusion Protein Ag17811
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
P glycoprotein Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated