missing translation for 'onlineSavingsMsg'
Learn More

p-Iodonitrotetrazolium Violet, MP Biomedicals

Product Code. p-7112287 Shop All MP Biomedical Products
100 mg, 100mg
Click to view available options
Quantity:
100 mg
500 mg
missing translation for 'unitSize'
100mg
500mg
This item is not returnable. View return policy
This item is not returnable. View return policy

  • p-Iodonitrotetazolium Violet is used as a electron acceptor for the colorimetric assay of various dehydrogenases

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146-68-9
Molecular Formula C19H13ClIN5O2
Molecular Weight (g/mol) 505.70
MDL Number MFCD00011961,MFCD00149999
InChI Key JORABGDXCIBAFL-UHFFFAOYSA-M
Synonym iodonitrotetrazolium chloride, iodonitrotetrazolium, iodonitrotetrazolium purple, iodonitrotetrazolium violet, 2-4-iodophenyl-3-4-nitrophenyl-5-phenyltetrazolium chloride, iodonitro tetrazolium, int, p-iodonitrotetrazolium violet, int chemical, 2-p-iodophenyl-3-p-nitrophenyl-5-phenyltetrazolium chloride
PubChem CID 64957
ChEBI CHEBI:75421
IUPAC Name 3-(4-iodophenyl)-2-(4-nitrophenyl)-5-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
SMILES [Cl-].[O-][N+](=O)C1=CC=C(C=C1)[N+]1=NC(=NN1C1=CC=C(I)C=C1)C1=CC=CC=C1

Specifications

Melting Point 245°C to 251°C (literature)
Color Yellow
Linear Formula C19H13ClIN5O2
Quantity 100 mg
Solubility Information Soluble in 5% methanol/water(1:1)(clear yellow); slightly soluble in water, alcohol, DMSO.
Formula Weight 505.7
Physical Form Powder
Chemical Name or Material p-IODONITROTETRAZOLIUM VIOLET
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
p-Iodonitrotetrazolium Violet, MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.