missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAFAH2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10085-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Has a marked selectivity for phospholipids with short acyl chains at the sn-2 position. May share a common physiologic function with the plasma-type enzyme.Specifications
PAFAH2 | |
Polyclonal | |
Unconjugated | |
PAFAH2 | |
HSD PLA2, PAFAH2, SD PLA2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100163, 313611, 5051 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P83006, Q8VDG7, Q99487 | |
PAFAH2 | |
PAFAH2 Fusion Protein Ag0124 | |
150 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |