missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAIP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | PAIP2 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16894814
|
Proteintech
15583-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16884814
|
Proteintech
15583-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
PAIP2 | |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
Q6AXZ0, Q9BPZ3, Q9D6V8 | |
361309, 51247, 67869 | |
PAIP2 Fusion Protein Ag7119 | |
Primary | |
-20°C | |
PAIP2 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
PABP interacting protein 2, PAIP 2, PAIP2, PAIP2A | |
Paip2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PAIP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech