missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAK4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14685-1-AP-20UL
This item is not returnable.
View return policy
Description
The p21-activated kinases (PAKs) are serine-threonine kinases that bind to the active forms of Cdc42 and Rac. They are divided into two groups, the first of which include PAK1, 2 and 3, and can be activated by Cdc42/Rac binding. The second group includes PAK4, 5 and 6. These proteins are not activated by Cdc42/Rac binding. PAK4 was initially identified as a novel effector of Cdc42Hs. Co-expression of PAK4 and Cdc42Hs resulted in induction of filopodia and actin polymerization, showing that it is involved in cytoskeletal reorganization. Other experiments have shown PAK4 to be essential for embryonic viability and proper neuronal development. PAK4 has also been implicated in anchorage-independent growth of tumor cells and is required for activation of several cancer prosurvival pathways.Specifications
PAK4 | |
Polyclonal | |
Unconjugated | |
PAK4 | |
KIAA1142, p21 activated kinase 4, PAK 4, PAK4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10298, 292756, 70584 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry, Immunoprecipitation, Immunocytochemistry | |
0.51 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O96013, Q8BTW9 | |
Pak4 | |
PAK4 Fusion Protein Ag5981 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PAK4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction