Learn More
PAPOLB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12821-1-AP-150UL
This item is not returnable.
View return policy
Description
Testis-specific poly(A) polymerase.Specifications
PAPOLB | |
Polyclonal | |
Unconjugated | |
PAPOLB | |
PAP beta, PAPOLB, PAPT, Poly(A) polymerase beta, TPAP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
304300, 56522, 56903 | |
-20°C | |
Liquid |
Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NRJ5, Q9WVP6 | |
Papolb | |
PAPOLB Fusion Protein Ag3487 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Product Title
PAPOLB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction