missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARP1 Mouse anti-Human, Mouse, Rat, Clone: 1D7D4, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | PARP1 |
---|---|
Clone | 1D7D4 |
Concentration | 0.5 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16828475
|
Proteintech
66520-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16818475
|
Proteintech
66520-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody was generated against the N-terminal region of human PARP1 and it recognizes the full-length as well as the cleavage of the PARP1.
Poly ADP-Ribose Polymerase (PARP) uses nicotinamide adenine dinucleotide (oxidized form) NAD as a substrate to catalyse the transfer of ADP-ribose to a variety of nuclear protein acceptors. Proteolysis of PARP to its stable 85kDa fragment is an early marker of programmed cell death (apoptosis) and is mediated by the caspase CPP32 protein. Cleavage occurs between Adp216 and Gly217, a site in PARP conserved across species.Specifications
PARP1 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ADPRT, ADPRT 1, ADPRT1, pADPRT 1, PARP, PARP 1, PARP1, poly (ADP ribose) polymerase 1, Poly [ADP ribose] polymerase 1, Poly[ADP ribose] synthase 1, PPOL | |
PARP1 | |
IgG1 | |
Protein G | |
Antibody |
1D7D4 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P09874, P11103, P27008 | |
11545, 142, 25591 | |
PARP1 Fusion Protein Ag19173 | |
Primary | |
-20°C | |
PARP1 |