Learn More
PARP10 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26072-1-AP-20UL
 View more versions of this product
Product Code. 16884595
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
E3 ubiquitin-protein ligase which is a component of the N-end rule pathway. Recognizes and binds to proteins bearing specific amino-terminal residues that are destabilizing according to the N-end rule, leading to their ubiquitination and subsequent degradation.
Specifications
Specifications
PARP10
Polyclonal
Unconjugated
PARP10
PARP 10, PARP10
Rabbit
Antigen Affinity Chromatography
RUO
84875
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation
0.39 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q53GL7
PARP10
PARP10 Fusion Protein Ag23457
20 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PARP10 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated