Learn More
CleanSpace Technology™ Particulate Pre-filter Product Code.: 15910062
Only undefined left
Add to basket
CleanSpace Technology™ Particulate Pre-filter
Product Code. 15910062 Shop All CleanSpace Technology Products
Pre-filter
Click to view available options
:
Pre-filter
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15910062
Brand: CleanSpace Technology™ PAF0036
Only undefined left
Add to basket
Only undefined left
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Suitable for use with the CleanSpace Standard Particulate P3 TM3 P SL R Filter for protection against airborne particulate (dust, mists and fumes). Designed to remove course particles and enhance the life of the Particulate P3 filter. The Pre-Filter should be changed after each use.
  • Used with the revolutionary CleanSpace - A light weight PAPR with no hoses/belts
  • Materials: Spun polymer fibers
  • Easy and quickly fitted and removed from the power unit
  • Sold in Pack of 10 pre-filters
  • Compatible with all CleanSpace Respirators
Specifications
Product Type Prefilter
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CleanSpace Technology™ Particulate Pre-filter >