missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 51036-2-AP-20UL
This item is not returnable.
View return policy
Description
Pax 3 (Paired Box 3) is a transcription factor that is involved in development of the peripheral nervous system, melanocytes, some vascular smooth muscle, and a number of other derivatives. It regulates neurogenesis in pre-migratory neural crest cells from the dorsal neural tube, and in myogenic progenitors in the presomitic mesoderm and the hypaxial somites. Alternative splicing results in isoforms having different C-termini.Specifications
PAX3 | |
Polyclonal | |
Unconjugated | |
PAX3 | |
CDHS, HUP2, paired box 3, Paired box protein Pax 3, PAX3, WS1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18505, 5077 | |
-20°C | |
Liquid |
Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P23760, P24610 | |
PAX3 | |
PAX3 Fusion Protein Ag0477 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PAX3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction