Learn More
PAX3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech 21386-1-AP-150UL
This item is not returnable.
View return policy
Beschrijving
Pax 3 (Paired Box 3) is a transcription factor that is involved in development of the peripheral nervous system, melanocytes, some vascular smooth muscle, and a number of other derivatives. It regulates neurogenesis in pre-migratory neural crest cells from the dorsal neural tube, and in myogenic progenitors in the presomitic mesoderm and the hypaxial somites. Alternative splicing results in isoforms having different C-termini.Specificatie
PAX3 | |
Polyclonal | |
Unconjugated | |
PAX3 | |
CDHS, HUP2, paired box 3, Paired box protein Pax 3, PAX3, WS1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114502, 18505, 5077 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.59 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P23760, P24610 | |
PAX3 | |
PAX3 Fusion Protein Ag16021 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Product Title
PAX3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction