Learn More
PAX3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21386-1-AP-150UL
 View more versions of this product
Product Code. 16880985
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
Pax 3 (Paired Box 3) is a transcription factor that is involved in development of the peripheral nervous system, melanocytes, some vascular smooth muscle, and a number of other derivatives. It regulates neurogenesis in pre-migratory neural crest cells from the dorsal neural tube, and in myogenic progenitors in the presomitic mesoderm and the hypaxial somites. Alternative splicing results in isoforms having different C-termini.
Specifications
Specifications
PAX3
Polyclonal
Unconjugated
PAX3
CDHS, HUP2, paired box 3, Paired box protein Pax 3, PAX3, WS1
Rabbit
Antigen Affinity Chromatography
RUO
114502, 18505, 5077
-20°C
Liquid
Immunoprecipitation, Western Blot
0.59 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P23760, P24610
PAX3
PAX3 Fusion Protein Ag16021
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PAX3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated