Learn More
PAX5 / BSAP (Early B-Cell Marker) Mouse, Clone: PCRP-PAX5-1B7, enQuireBio™
Mouse Monoclonal Antibody
2050.00 SEK - 5965.00 SEK
Specifications
Antigen | PAX5 / BSAP (Early B-Cell Marker) |
---|---|
Clone | PCRP-PAX5-1B7 |
Concentration | 200ÎĽg/mL |
Applications | Immunoprecipitation, Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15910608
|
EnquireBio
5079-MSM1-P0 |
20 ÎĽg | N/A | ||||||
15920608
|
EnquireBio
5079-MSM1-P1 |
100 ÎĽg |
5965.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
PAX5 / BSAP (Early B-Cell Marker) | |
200ÎĽg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human PAX5 protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
42kDa |
PCRP-PAX5-1B7 | |
Immunoprecipitation, Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
5079 | |
IgG1 | |
Protein A/G purified from Bioreactor concentrate. | |
Nucleus | |
Raji, DAUDI or Ramos Cells. Tonsil, spleen, Lymph Node or Colon Carcinoma. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
PAX5 / BSAP (Early B-Cell Marker) Mouse, Clone: PCRP-PAX5-1B7, enQuireBio™