missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Pax6 Mouse, Clone: PAX6/496, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-32893-0.02mg
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Pax6 Monoclonal specifically detects Pax6 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).
Specifikationer
| Pax6 | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| keratitis), MGC17209, Oculorhombin, paired box 6, paired box protein Pax-6 | |
| Mouse | |
| 47 kDa | |
| 0.02 mg | |
| Cellular Markers, Diabetes Research, Neuronal Stem Cell Markers, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision | |
| 5080 | |
| Human, Mouse, Rat, Chicken | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| PAX6/496 | |
| Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml | |
| P26367 | |
| PAX6 | |
| Recombinant fragment (N-terminus; aa 1-300) of human PAX6 protein | |
| Protein A purified | |
| RUO | |
| Primary | |
| Pax genes contain paired domains with strong homology to genes in Drosophila, which are involved in programming early development. Lesions in the Pax-6 gene account for most cases of aniridia, a congenital malformation of the eye, chiefly characterized by iris hypoplasia, which can cause blindness. Pax-6 is involved in other anterior segment malformations besides aniridia, such as Peters anomaly, a major error in the embryonic development of the eye with corneal clouding with variable iridolenticulocorneal adhesions. The Pax-6 gene encodes a transcriptional regulator that recognizes target genes through its paired-type DNA-binding domain. The paired domain is composed of two distinct DNA-binding subdomains, the amino-terminal subdomain and the carboxy-terminal subdomain, which bind respective consensus DNA sequences. The human Pax-6 gene produces two alternatively spliced isoforms that have the distinct structure of the paired domain. | |
| Store at 4C. | |
| IgG1 κ |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering