missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Pax6 Mouse, Clone: PAX6/496, Novus Biologicals™
Mouse Monoclonal Antibody
2815.00 SEK - 4880.00 SEK
Specifikationer
| Antigen | Pax6 |
|---|---|
| Klona | PAX6/496 |
| Utspädning | Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml |
| Användningsområden | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Klassificering | Monoclonal |
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|
18761774
|
Novus Biologicals
NBP2-32893-0.02mg |
0.02 mg |
2815.00 SEK
0.02mg |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
|
18186054
|
Novus Biologicals
NBP2-32893-0.1MG |
0.1 mg |
4880.00 SEK
0.10mg |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
Beskrivning
Pax6 Monoclonal specifically detects Pax6 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).Specifikationer
| Pax6 | |
| Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Chicken | |
| P26367 | |
| 5080 | |
| Recombinant fragment (N-terminus; aa 1-300) of human PAX6 protein | |
| Primary | |
| Store at 4C. | |
| 47 kDa |
| PAX6/496 | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cellular Markers, Diabetes Research, Neuronal Stem Cell Markers, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| keratitis), MGC17209, Oculorhombin, paired box 6, paired box protein Pax-6 | |
| PAX6 | |
| IgG1 κ | |
| Protein A purified | |
| Pax genes contain paired domains with strong homology to genes in Drosophila, which are involved in programming early development. Lesions in the Pax-6 gene account for most cases of aniridia, a congenital malformation of the eye, chiefly characterized by iris hypoplasia, which can cause blindness. Pax-6 is involved in other anterior segment malformations besides aniridia, such as Peters anomaly, a major error in the embryonic development of the eye with corneal clouding with variable iridolenticulocorneal adhesions. The Pax-6 gene encodes a transcriptional regulator that recognizes target genes through its paired-type DNA-binding domain. The paired domain is composed of two distinct DNA-binding subdomains, the amino-terminal subdomain and the carboxy-terminal subdomain, which bind respective consensus DNA sequences. The human Pax-6 gene produces two alternatively spliced isoforms that have the distinct structure of the paired domain. |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel