missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDHGB3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21204-1-AP-150UL
This item is not returnable.
View return policy
Specifications
PCDHGB3 | |
Polyclonal | |
Unconjugated | |
PCDHGB3 | |
PCDH GAMMA B3, PCDHGB3, Protocadherin gamma B3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56102 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Y5G1 | |
PCDHGB3 | |
PCDHGB3 Fusion Protein Ag15569 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PCDHGB3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction