Learn More
MP Biomedicals™ PCR buffer 10x without MGCl2 (5 x 1mL)
Brand:  MP Biomedicals™ 11EPTQBUF2
Product Code. 11437020
  • 595.00 SEK / 5mL
Specifications
Store at -20°C.
5x 1mL
Certificates
Promotions
Promotions
Product Content Correction
Product Title
MP Biomedicals™ PCR buffer 10x without MGCl2 (5 x 1mL) >