missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCYT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14827-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes an enzyme that catalyzes the formation of CDP-ethanolamine from CTP and phosphoethanolamine in the Kennedy pathway of phospholipid synthesis. Alternative splicing results in multiple transcript variants.Specifications
PCYT2 | |
Polyclonal | |
Unconjugated | |
PCYT2 | |
ET, PCYT2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5833, 68671, 89841 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88637, Q922E4, Q99447 | |
PCYT2 | |
PCYT2 Fusion Protein Ag6616 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PCYT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction