Läs mer
Tocris Bioscience™ PD 173074
GREENER_CHOICE
Produktkod. 18064312
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18064312 10 mg
18044882 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18064312 Leverantör Tocris Bioscience™ Leverantörsnummer 3044/10
FGFR1 and -3 inhibitor
PD 173074 is a selective FGFR1 and FGFR3 inhibitor (IC50 values are 5, 21.5, ∼100, 17600 and 19800 nM for FGFR3, FGFR1, VEGFR2, PDGFR and c-Src respectively, and > 50000 nM for EGFR, InsR, MEK and PKC). Inhibits VEGF- and FGF-induced angiogenesis in the mouse cornea model of angiogenesis. Inhibits proliferation and differentiation of oligodendrocyte progenitors. Also promotes mESC self-renewal, facilitates the conversion of mouse epiblast stem cells to an earlier pluripotency state and inhibits differentiation of miPSCs to cardiomyocytes. Suppresses cell proliferation in cell lines expressing mutated FGFR3 protein. Blocks tumor growth in H510 and H69 SCLC xenograft models.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 219580-11-7
Molekylformel C28H41N7O3
Specifikationer
Kemiskt namn eller material PD 173074
Kvantitet 10 mg
Formel vikt 523.67
Renhet >98%
Molekylvikt (g/mol) 523.682