missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-ECGF Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | PD-ECGF |
---|---|
Concentration | 0.21 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890074
|
Proteintech
12383-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880074
|
Proteintech
12383-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Platelet - derived endothelial growth factor (PD-ECGF), same as thymidine phosphorylase (TP) or gliostatin. In the presence of inorganic orthophosphate, it catalyses the reversible phospholytic cleavage of thymidine and deoxyuridine to their correspondingbases and 2-deoxyribose-1-phosphate. It is both chemotactic and mitogenic for endothelial cells and a non-heparin binding angiogenic factor present in platelets. It is also invoved in transformation of fluoropyrimidines, cytotoxic agents used in the treatment of a variety of maligcies, into active cytotoxic metabolites.Specifications
PD-ECGF | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P19971 | |
1890 | |
PD-ECGF Fusion Protein Ag3052 | |
Primary | |
-20°C | |
TYMP |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ECGF1, Gliostatin, hPD ECGF, MNGIE, PD ECGF, PDECGF, TdRPase, thymidine phosphorylase, TP, TYMP | |
TYMP | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PD-ECGF Rabbit anti-Human, Polyclonal, Proteintech